2-(Benzyloxy)-5-chloro-1,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

64832-500MG 64832-1G 64832-5G 64832-25G
77332-342EA 136.99 USD
77332-342 77344-560 77368-406 77389-676
2-(Benzyloxy)-5-chloro-1,3-difluorobenzene ≥97%
2-(Benzyloxy)-5-chloro-1,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉ClF₂O
MW: 254.6597 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044307
CAS Number: 1881296-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR