3,5-Dibromo-6-chloropyridin-2-ol ≥95%

Supplier: AOBChem USA

Warning

64820-1G 64820-5G 64820-25G
77340-878EA 61.8 USD
77340-878 77368-402 77380-602
3,5-Dibromo-6-chloropyridin-2-ol ≥95%
3,5-Dibromo-6-chloropyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₂Br₂ClNO
MW: 287.3364 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015847
CAS Number: 6515-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR