2,3,5-Tribromo-6-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

64819-500MG 64819-1G 64819-5G 64819-25G
77332-338EA 109.18 USD
77332-338 77343-224 77368-400 77380-600
2,3,5-Tribromo-6-chloropyridine ≥97%
2,3,5-Tribromo-6-chloropyridine

Precaución: for Research Use Only

Formula: C₅HBr₃ClN
Storage Temperature: Ambient
MDL Number: MFCD34761984
CAS Number: 2807467-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR