3,5-Dibromo-2-chloro-4-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64816-500MG 64816-1G 64816-5G 64816-25G
77332-336EA 252.35 USD
77332-336 77356-656 77368-398 77389-672
3,5-Dibromo-2-chloro-4-iodobenzoic acid ≥97%
3,5-Dibromo-2-chloro-4-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂Br₂ClIO₂
Storage Temperature: Ambient
MDL Number: MFCD34761983
CAS Number: 2807464-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR