3,4,5-Tribromo-2-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64813-500MG 64813-1G 64813-5G
77332-334EA 328.57 USD
77332-334 77356-654 77380-598
3,4,5-Tribromo-2-chlorobenzoic acid ≥97%
3,4,5-Tribromo-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂Br₃ClO₂
MW: 393.2545 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761982
CAS Number: 2807443-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR