(4-Fluoro-[1,1'-biphenyl]-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

64804-500MG 64804-1G 64804-5G
77335-886EA 367.71 USD
77335-886 77356-652 77380-596
(4-Fluoro-[1,1'-biphenyl]-3-yl)methanol ≥97%
(4-Fluoro-[1,1'-biphenyl]-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₁FO
MW: 202.2242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701923
CAS Number: 89951-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR