3,5-Dibromo-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64796-500MG 64796-1G 64796-5G 64796-25G
77332-330EA 82.4 USD
77332-330 77342-162 77368-394 77380-594
3,5-Dibromo-2-methoxybenzaldehyde ≥97%
3,5-Dibromo-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MW: 293.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093682
CAS Number: 61657-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR