5-Bromo-3-chloro-2-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64788-500MG 64788-1G 64788-5G 64788-25G
77332-326EA 194.67 USD
77332-326 77356-650 77368-390 77389-666
5-Bromo-3-chloro-2-ethoxybenzaldehyde ≥97%
5-Bromo-3-chloro-2-ethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MW: 263.5156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761975
CAS Number: 2384593-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR