1-Chloro-2-ethoxy-3-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

64782-500MG 64782-1G 64782-5G 64782-25G
77332-322EA 135.96 USD
77332-322 77344-516 77368-386 77389-662
1-Chloro-2-ethoxy-3-iodobenzene ≥95%
1-Chloro-2-ethoxy-3-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClIO
MW: 282.5059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761974
CAS Number: 2332809-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR