(3-Bromo-2-fluoro-5-isopropylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

64743-500MG 64743-1G 64743-5G
77338-940EA 532.51 USD
77338-940 77356-648 77380-588
(3-Bromo-2-fluoro-5-isopropylphenyl)(methyl)sulfane ≥95%
(3-Bromo-2-fluoro-5-isopropylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFS
Storage Temperature: Ambient
MDL Number: MFCD34761947
CAS Number: 2807446-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR