3-Bromo-2-fluoro-5-isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64740-500MG 64740-1G 64740-5G
77337-206EA 408.91 USD
77337-206 77356-644 77380-584
3-Bromo-2-fluoro-5-isopropylbenzaldehyde ≥97%
3-Bromo-2-fluoro-5-isopropylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO
MW: 245.0882 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761944
CAS Number: 1781010-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR