3,5-Dichloro-4-iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

64734-500MG 64734-1G 64734-5G 64734-25G
77332-312EA 126.69 USD
77332-312 77344-188 77368-376 77380-582
3,5-Dichloro-4-iodo-2-methoxypyridine ≥97%
3,5-Dichloro-4-iodo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂INO
MW: 303.9125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32061849
CAS Number: 1900707-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR