3,5-Dichloro-2-methoxyisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

64731-500MG 64731-1G 64731-5G 64731-25G
77332-310EA 209.09 USD
77332-310 77356-640 77368-374 77389-656
3,5-Dichloro-2-methoxyisonicotinic acid ≥95%
3,5-Dichloro-2-methoxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₃
MW: 222.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025889
CAS Number: 1147979-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR