4-Fluoro-3-iodo-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64715-500MG 64715-1G 64715-5G 64715-25G
77332-306EA 120.51 USD
77332-306 77343-782 77368-370 77380-580
4-Fluoro-3-iodo-5-methoxybenzaldehyde ≥97%
4-Fluoro-3-iodo-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
Storage Temperature: Ambient
MDL Number: MFCD34761929
CAS Number: 2384860-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR