1-Fluoro-2-iodo-4-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

64705-500MG 64705-1G 64705-5G
77338-728EA 511.91 USD
77338-728 77356-636 77380-576
1-Fluoro-2-iodo-4-isopropylbenzene ≥97%
1-Fluoro-2-iodo-4-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FI
Storage Temperature: Ambient
MDL Number: MFCD29091189
CAS Number: 1369776-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR