1,3-Difluoro-2-isopropoxy-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

64688-500MG 64688-1G 64688-5G 64688-25G
77332-298EA 110.21 USD
77332-298 77343-276 77368-362 77380-570
1,3-Difluoro-2-isopropoxy-4-methoxybenzene ≥97%
1,3-Difluoro-2-isopropoxy-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
MW: 202.1978 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761912
CAS Number: 2807450-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR