2-(Benzyloxy)-1,3-difluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

64687-1G 64687-5G 64687-25G
77341-488EA 89.61 USD
77341-488 77368-360 77380-568
2-(Benzyloxy)-1,3-difluoro-4-methoxybenzene ≥97%
2-(Benzyloxy)-1,3-difluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂O₂
MW: 250.2406 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761911
CAS Number: 2807463-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR