5-Bromo-2-chloro-1-fluoro-3-methylbenzene ≥95%
Supplier: AOBChem USA
|
Warning
|
64672-500MG
64672-1G
64672-5G
77332-294EA
322.39
USD
77332-294
77356-630
77380-564
5-Bromo-2-chloro-1-fluoro-3-methylbenzene ≥95%
5-Bromo-2-chloro-1-fluoro-3-methylbenzene
Precaución: for Research Use Only
Formula:
C₇H₅BrClF MW: 223.4700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28734050 CAS Number: 1806971-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...