1-Bromo-2-fluoro-3-iodo-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

64663-500MG 64663-1G 64663-5G 64663-25G
77332-290EA 168.92 USD
77332-290 77346-304 77368-356 77389-646
1-Bromo-2-fluoro-3-iodo-4-methylbenzene ≥95%
1-Bromo-2-fluoro-3-iodo-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
MW: 314.9214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368769
CAS Number: 886762-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR