(2-Fluoro-3-iodo-6-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

64662-500MG 64662-1G 64662-5G 64662-25G
77332-288EA 161.71 USD
77332-288 77345-932 77368-354 77389-644
(2-Fluoro-3-iodo-6-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-iodo-6-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈FIS
Storage Temperature: Ambient
MDL Number: MFCD34761900
CAS Number: 2705797-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR