(2-Fluoro-3-iodo-5-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

64653-500MG 64653-1G 64653-5G 64653-25G
77332-280EA 184.37 USD
77332-280 77347-266 77368-348 77389-638
(2-Fluoro-3-iodo-5-methoxyphenyl)(methyl)sulfane ≥95%
(2-Fluoro-3-iodo-5-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈FIOS
Storage Temperature: Ambient
MDL Number: MFCD34761894
CAS Number: 2705647-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR