(2-Fluoro-5-methoxy-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

64652-500MG 64652-1G 64652-5G 64652-25G
77332-278EA 182.31 USD
77332-278 77346-990 77368-346 77389-636
(2-Fluoro-5-methoxy-1,3-phenylene)bis(methylsulfane) ≥97%
(2-Fluoro-5-methoxy-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₉H₁₁FOS₂
MW: 218.3114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761893
CAS Number: 2807468-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR