2-Fluoro-5-methoxy-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64650-500MG 64650-1G 64650-5G 64650-25G
77332-276EA 182.31 USD
77332-276 77346-988 77368-344 77389-634
2-Fluoro-5-methoxy-3-(methylthio)benzaldehyde ≥97%
2-Fluoro-5-methoxy-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
MW: 200.2299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761891
CAS Number: 2091772-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR