1-Fluoro-4-methoxy-2-(methylsulfonyl)benzene ≥95%

Supplier: AOBChem USA

Warning

64645-1G 64645-5G 64645-25G
77341-952EA 108.15 USD
77341-952 77368-338 77380-558
1-Fluoro-4-methoxy-2-(methylsulfonyl)benzene ≥95%
1-Fluoro-4-methoxy-2-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₉FO₃S
MW: 204.2186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761887
CAS Number: 652171-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR