5-Ethoxy-2-fluoro-3-(methylthio)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

64632-500MG 64632-1G 64632-5G 64632-25G
77332-266EA 184.37 USD
77332-266 77347-264 77368-334 77389-626
5-Ethoxy-2-fluoro-3-(methylthio)benzoic acid ≥95%
5-Ethoxy-2-fluoro-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃S
Storage Temperature: Ambient
MDL Number: MFCD34761875
CAS Number: 2807455-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR