4'-Methyl-2-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64598-500MG 64598-1G 64598-5G 64598-25G
77332-258EA 111.24 USD
77332-258 77343-290 77368-322 77380-546
4'-Methyl-2-nitro-1,1'-biphenyl ≥97%
4'-Methyl-2-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂
MW: 213.2319 g/mol
Storage Temperature: Ambient
CAS Number: 70680-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR