3-(2,5-Dibromophenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

64597-500MG 64597-1G 64597-5G 64597-25G
77332-256EA 141.11 USD
77332-256 77344-858 77368-320 77389-618
3-(2,5-Dibromophenyl)pyridine ≥95%
3-(2,5-Dibromophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇Br₂N
Storage Temperature: Ambient
MDL Number: MFCD34761856
CAS Number: 2677851-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR