4-(2,5-Dibromophenyl)pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
64596-500MG
64596-1G
64596-5G
64596-25G
77332-254EA
116.39
USD
77332-254
77343-442
77368-318
77380-544
4-(2,5-Dibromophenyl)pyridine ≥97%
4-(2,5-Dibromophenyl)pyridine
Precaución: for Research Use Only
Formula:
C₁₁H₇Br₂N Storage Temperature: Ambient |
MDL Number:
MFCD34761855 CAS Number: 2807467-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...