2,5-Dibromo-4'-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64595-500MG 64595-1G 64595-5G 64595-25G
77332-252EA 99.91 USD
77332-252 77342-854 77368-316 77380-542
2,5-Dibromo-4'-methoxy-1,1'-biphenyl ≥97%
2,5-Dibromo-4'-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀Br₂O
MW: 342.0259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761854
CAS Number: 66175-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR