4-(4-Bromo-3-(methylthio)phenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

64589-500MG 64589-1G 64589-5G
77336-200EA 381.1 USD
77336-200 77356-608 77380-540
4-(4-Bromo-3-(methylthio)phenyl)pyridine ≥95%
4-(4-Bromo-3-(methylthio)phenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNS
MW: 280.1835 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761849
CAS Number: 2901097-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR