(4-Bromo-4'-methyl-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

64584-500MG 64584-1G 64584-5G
77332-246EA 323.42 USD
77332-246 77356-604 77380-536
(4-Bromo-4'-methyl-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%
(4-Bromo-4'-methyl-[1,1'-biphenyl]-3-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrS
MW: 293.2220 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761844
CAS Number: 2901102-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR