(4-Bromo-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

64578-500MG 64578-1G 64578-5G 64578-25G
77332-244EA 264.71 USD
77332-244 77356-602 77368-312 77389-614
(4-Bromo-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥95%
(4-Bromo-[1,1'-biphenyl]-3-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrS
Storage Temperature: Ambient
MDL Number: MFCD34761840
CAS Number: 2901109-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR