5-Bromo-1-ethyl-2-fluoro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

64567-500MG 64567-1G 64567-5G
77335-884EA 367.71 USD
77335-884 77356-596 77380-530
5-Bromo-1-ethyl-2-fluoro-3-methylbenzene ≥95%
5-Bromo-1-ethyl-2-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrF
MW: 217.0781 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761831
CAS Number: 2710571-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR