(2-(Benzyloxy)-6-chlorophenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

64563-500MG 64563-1G 64563-5G
77332-240EA 350.2 USD
77332-240 77356-594 77380-528
(2-(Benzyloxy)-6-chlorophenyl)boronic acid ≥95%
(2-(Benzyloxy)-6-chlorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClO₃
Storage Temperature: Ambient
MDL Number: MFCD34761828
CAS Number: 1887046-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR