(2,3-Dichloro-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

64559-500MG 64559-1G 64559-5G
77338-400EA 485.13 USD
77338-400 77356-590 77380-524
(2,3-Dichloro-4-methylphenyl)(methyl)sulfane ≥97%
(2,3-Dichloro-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂S
MW: 207.1201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28787035
CAS Number: 1803813-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR