1-(2,3-Dichloro-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

64557-500MG 64557-1G 64557-5G
77337-984EA 456.29 USD
77337-984 77356-588 77380-522
1-(2,3-Dichloro-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,3-Dichloro-4-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅Cl₂F₃O
MW: 257.0366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761825
CAS Number: 2901098-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR