(2-(Benzyloxy)-3-fluoro-4-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

64543-500MG 64543-1G 64543-5G
77337-940EA 453.2 USD
77337-940 77356-586 77380-520
(2-(Benzyloxy)-3-fluoro-4-methylphenyl)(methyl)sulfane ≥95%
(2-(Benzyloxy)-3-fluoro-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₅H₁₅FOS
MW: 262.3424 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761814
CAS Number: 2901098-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR