2-(Benzyloxy)-5-bromo-1-chloro-3-fluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

64515-500MG 64515-1G 64515-5G 64515-25G
77332-234EA 202.91 USD
77332-234 77356-582 77368-306 77389-606
2-(Benzyloxy)-5-bromo-1-chloro-3-fluoro-4-methylbenzene ≥97%
2-(Benzyloxy)-5-bromo-1-chloro-3-fluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34761811
CAS Number: 2918853-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR