1-Bromo-5-chloro-3-fluoro-4-isopropoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

64514-500MG 64514-1G 64514-5G 64514-25G
77332-232EA 95.79 USD
77332-232 77342-578 77368-304 77380-516
1-Bromo-5-chloro-3-fluoro-4-isopropoxy-2-methylbenzene ≥95%
1-Bromo-5-chloro-3-fluoro-4-isopropoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34761810
CAS Number: 2807455-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR