1-Bromo-5-chloro-4-ethoxy-3-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

64513-500MG 64513-1G 64513-5G 64513-25G
77332-230EA 143.17 USD
77332-230 77345-004 77368-302 77389-604
1-Bromo-5-chloro-4-ethoxy-3-fluoro-2-methylbenzene ≥97%
1-Bromo-5-chloro-4-ethoxy-3-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34761809
CAS Number: 2807443-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR