2-(Benzyloxy)-1-chloro-3-fluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

64510-500MG 64510-1G 64510-5G 64510-25G
77332-226EA 105.06 USD
77332-226 77343-096 77368-298 77380-512
2-(Benzyloxy)-1-chloro-3-fluoro-4-methylbenzene ≥97%
2-(Benzyloxy)-1-chloro-3-fluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFO
MW: 250.6958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761806
CAS Number: 2807467-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR