(4-Ethoxy-2-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

64509-1G 64509-5G 64509-25G
77341-352EA 82.4 USD
77341-352 77368-296 77380-510
(4-Ethoxy-2-fluorophenyl)(methyl)sulfane ≥97%
(4-Ethoxy-2-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD34761805
CAS Number: 2901106-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR