4-Bromo-4'-methoxy-2-(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64505-500MG 64505-1G 64505-5G 64505-25G
77332-224EA 204.97 USD
77332-224 77356-580 77368-294 77389-602
4-Bromo-4'-methoxy-2-(trifluoromethoxy)-1,1'-biphenyl ≥97%
4-Bromo-4'-methoxy-2-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O₂
MW: 347.1272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761801
CAS Number: 2807444-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR