4-Bromo-4'-fluoro-2-(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64504-500MG 64504-1G 64504-5G 64504-25G
77332-222EA 161.71 USD
77332-222 77345-928 77368-292 77389-600
4-Bromo-4'-fluoro-2-(trifluoromethoxy)-1,1'-biphenyl ≥97%
4-Bromo-4'-fluoro-2-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₄O
MW: 335.0916 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761800
CAS Number: 2807454-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR