4-Bromo-4'-methyl-2-(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64502-500MG 64502-1G 64502-5G 64502-25G
77332-220EA 152.44 USD
77332-220 77345-486 77368-290 77389-598
4-Bromo-4'-methyl-2-(trifluoromethoxy)-1,1'-biphenyl ≥97%
4-Bromo-4'-methyl-2-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O
MW: 331.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761798
CAS Number: 2807441-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR