4-Bromo-2-(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64499-500MG 64499-1G 64499-5G 64499-25G
77332-218EA 147.29 USD
77332-218 77345-240 77368-288 77389-596
4-Bromo-2-(trifluoromethoxy)-1,1'-biphenyl ≥97%
4-Bromo-2-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₈BrF₃O
MW: 317.1012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761796
CAS Number: 2807450-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR