2-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

64489-500MG 64489-1G 64489-5G
77339-066EA 547.96 USD
77339-066 77356-576 77380-506
2-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid ≥97%
2-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O₂
MW: 266.2152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702290
CAS Number: 916806-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR