2-Chloro-4-fluoro-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64477-500MG 64477-1G 64477-5G
77335-606EA 360.5 USD
77335-606 77356-574 77380-504
2-Chloro-4-fluoro-4'-methyl-1,1'-biphenyl ≥97%
2-Chloro-4-fluoro-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClF
MW: 220.6699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761783
CAS Number: 2807447-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR