2-Chloro-4-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64476-500MG 64476-1G 64476-5G 64476-25G
77332-212EA 264.71 USD
77332-212 77356-572 77368-282 77389-590
2-Chloro-4-fluoro-1,1'-biphenyl ≥97%
2-Chloro-4-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈ClF
MW: 206.6433 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761782
CAS Number: 39224-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR