7-(Methylthio)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

64463-500MG 64463-1G
77340-522EA 1437.88 USD
77340-522 77356-570
7-(Methylthio)-1H-indazole ≥97%
7-(Methylthio)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂S
MW: 164.2275 g/mol
Storage Temperature: Ambient
CAS Number: 2243713-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR